Abnova
Manufacturer Code:PAB22683
Catalog # 89-124-391
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human MTHFD2L. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant MTHFD2L.
For Research Use Only
Protein Aliases: FLJ13105; Methenyltetrahydrofolate cyclohydrolase; MGC45532; MGC72244; MTHFD2-like; NAD-dependent methylenetetrahydrofolate dehydrogenase; NADP-dependent methylenetetrahydrofolate dehydrogenase 2-like protein; Probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; tetrahydrofolate dehydrogenase/cyclohydrolase
Gene Aliases: MTHFD2L
UniProt ID: (Human) Q9H903
Entrez Gene ID: (Human) 441024
Molecular Function:
dehydrogenase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.