Novus Biologicals
Manufacturer Code:NBP297866R
Catalog # NB048710
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human MTHFD2 fragment. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase methenyltetrahydrofolate cyclohydrolase methylene tetrahydrofolate dehydrogenase (NAD+ dependent) methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2 mitochondrial NAD-dependent methylene tetrahydrofolate dehydrogenase cyclohydrolase NMDMC; Bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase, mitochondrial; Methenyltetrahydrofolate cyclohydrolase; NAD-dependent methylene tetrahydrofolate dehydrogenase cyclohydrolase; NAD-dependent methylenetetrahydrofolate dehydrogenase
Gene Aliases: MTHFD2; NMDMC
UniProt ID: (Human) P13995
Entrez Gene ID: (Human) 10797
Molecular Function: dehydrogenase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.