Novus Biologicals
Manufacturer Code:NBP237864
Catalog # NBP237864
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: HFGGLIEEDDVILLAAALRIQNMVSSGRRWLREQQHRRWRLHCLKLQPLSPVPSDIEISRGQTPKAVDVLAKEIGLLADEIEIYGKSKAKV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 10-formyl-THF synthetase; 10-formyl-THF synthetase dJ292B18.2 DKFZp586G1517 EC 6.3.4.3 FLJ21145 Formyltetrahydrofolate synthetase formyltetrahydrofolate synthetase domain containing 1 FTHFSDC1 methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like mitochondrial C1-tetrahydrofolate synthase mitochondrial C1-tetrahydrofolate synthetase monofunctional C1-tetrahydrofolate synthase mitochondrial MTC1THFS; Formyltetrahydrofolate synthetase; formyltetrahydrofolate synthetase domain containing 1; Monofunctional C1-tetrahydrofolate synthase, mitochondrial
Gene Aliases: dJ292B18.2; FTHFSDC1; MTC1THFS; MTHFD1L
UniProt ID: (Human) Q9UFU8
Entrez Gene ID: (Human) 25902
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.