Invitrogen
Manufacturer Code:PA542826
Catalog # PIPA542826
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human MTHFD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5,10-methylenetetrahydrofolate dehydrogenase, 5,10-methylenetetrahydrofolate cyclohydrolase, 10-formyltetrahydrofolate synthetase; C-1-tetrahydrofolate synthase, cytoplasmic; C-1-tetrahydrofolate synthase, cytoplasmic, N-terminally processed; C1-tetrahydrofolate synthase; C1-THF synthase; cytoplasmic C-1-tetrahydrofolate synthase; Formyltetrahydrofolate synthetase; Methenyltetrahydrofolate cyclohydrolase; Methylenetetrahydrofolate dehydrogenase; Methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1 methenyltetrahydrofolate cyclohydrolase formyltetrahydrofolate synthetase MTHFC MTHFD; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1, methenyltetrahydrofolate cyclohydrolase, formyltetrahydrofolate synthetase; methylenetetrahydrofolate dehydrogenase (NADP+ dependent), methenyltetrahydrofolate cyclohydrolase, formyltetrahydrofolate synthase; methylenetetrahydrofolate dehydrogenase 1; MTHFD1 protein; formyltetrahydrofolate synthetase; methenyltetrahydrofolate cyclohydrolase,
Gene Aliases: Dcs; E430024A07Rik; MTHFC; MTHFD; MTHFD1
UniProt ID: (Human) P11586, (Mouse) Q922D8, (Rat) P27653
Entrez Gene ID: (Rabbit) 100347271, (Pig) 414382, (Horse) 100062154, (Human) 4522, (Dog) 480352, (Bovine) 534382, (Mouse) 108156, (Guinea pig) 100734825, (Rat) 64300
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.