Novus Biologicals
Manufacturer Code:NBP106054
Catalog # NBP106054
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence RGDLNDCFIPCTPK corresponding to internal region according to NP_005947.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 5,10-methylenetetrahydrofolate dehydrogenase, 5,10-methylenetetrahydrofolate cyclohydrolase, 10-formyltetrahydrofolate synthetase; C-1-tetrahydrofolate synthase, cytoplasmic; C-1-tetrahydrofolate synthase, cytoplasmic, N-terminally processed; C1-THF synthase; C1-THF synthase EC 6.3.4.3 methenyltetrahydrofolate cyclohydrolase formyltetrahydrofolate synthetase510-methylenetetrahydrofolate dehydrogenase 510-methylenetetrahydrofolatecyclohydrolase 10-formyltetrahydrofolate synthetase methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1 MTHFCC-1-tetrahydrofolate synthase cytoplasmic MTHFDcytoplasmic C-1-tetrahydrofolate synthase; cytoplasmic C-1-tetrahydrofolate synthase; Formyltetrahydrofolate synthetase; Methenyltetrahydrofolate cyclohydrolase; Methylenetetrahydrofolate dehydrogenase; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1, methenyltetrahydrofolate cyclohydrolase, formyltetrahydrofolate synthetase
Gene Aliases: MTHFC; MTHFD; MTHFD1
UniProt ID: (Human) P11586
Entrez Gene ID: (Human) 4522
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.