Proteintech
Manufacturer Code:107941AP150UL
Catalog # 50-172-6604
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MTHFD1 Fusion Protein Ag1225 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5,10-methylenetetrahydrofolate dehydrogenase, 5,10-methylenetetrahydrofolate cyclohydrolase, 10-formyltetrahydrofolate synthetase; C-1-tetrahydrofolate synthase, cytoplasmic; C-1-tetrahydrofolate synthase, cytoplasmic, N-terminally processed; C1-tetrahydrofolate synthase; C1-THF synthase; cytoplasmic C-1-tetrahydrofolate synthase; Formyltetrahydrofolate synthetase; Methenyltetrahydrofolate cyclohydrolase; Methylenetetrahydrofolate dehydrogenase; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1, methenyltetrahydrofolate cyclohydrolase, formyltetrahydrofolate synthetase; MTHFC MTHFD MTHFD1
Gene Aliases: Dcs; E430024A07Rik; MTHFC; MTHFD; MTHFD1
UniProt ID: (Human) P11586, (Mouse) Q922D8
Entrez Gene ID: (Human) 4522, (Mouse) 108156
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.