Novus Biologicals
Manufacturer Code:NBP261160100UL
Catalog # NB261160100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AT3B3 |
Immunogen | Recombinant human NUDT1 (1-156aa) purified from E. coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: 2-hydroxy-dATP diphosphatase; 7,8-dihydro-8-oxoguanine triphosphatase; 8-oxo-7,8-dihydrodeoxyguanosine triphosphatase; 8-oxo-7,8-dihydroguanosine triphosphatase; 8-oxo-78-dihydrodeoxyguanosine triphosphatase EC 3.6.1.- MTH1Nucleoside diphosphate-linked moiety X motif 1 mutT human homolog 18-oxo-dGTPase nucleoside diphosphate-linked moiety X-type motif 178-dihydro-8-oxoguanine triphosphatase nudix (nucleoside diphosphate linked moiety X)-type motif 18-oxo-78-dihydroguanosine triphosphatase Nudix motif 1; 8-oxo-dGTPase; mutT human homolog 1; Nucleoside diphosphate-linked moiety X motif 1; nucleoside diphosphate-linked moiety X-type motif 1; nudix (nucleoside diphosphate linked moiety X)-type motif 1; Nudix motif 1
Gene Aliases: MTH1; NUDT1
UniProt ID: (Human) P36639
Entrez Gene ID: (Human) 4521
Molecular Function:
hydrolase
phosphorylase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.