Invitrogen
Manufacturer Code:MA535421
Catalog # PIMA535421
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0869 |
Immunogen | A synthesized peptide derived from human MTH1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 2-hydroxy-dATP diphosphatase; 2-hydroxy-dATP diphosphatase 78-dihydro-8-oxoguanine triphosphatase 8-oxo-dGTPase Mth1 Nucleoside diphosphate-linked moiety X motif 1 Nudix motif 1 NUDT1; 7,8-dihydro-8-oxoguanine triphosphatase; 8-oxo-7,8-dihydrodeoxyguanosine triphosphatase; 8-oxo-7,8-dihydroguanosine triphosphatase; 8-oxo-dGTPase; mutT human homolog 1; Nucleoside diphosphate-linked moiety X motif 1; nucleoside diphosphate-linked moiety X-type motif 1; nudix (nucleoside diphosphate linked moiety X)-type motif 1; Nudix motif 1
Gene Aliases: MTH1; NUDT1
UniProt ID: (Human) P36639
Entrez Gene ID: (Human) 4521
Molecular Function:
hydrolase
phosphorylase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.