Invitrogen
Manufacturer Code:PA5102780
Catalog # PIPA5102780
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NUDT1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-hydroxy-dATP diphosphatase; 2-hydroxy-dATP diphosphatase 78-dihydro-8-oxoguanine triphosphatase 8-oxo-78-dihydro-2'-dGTPase 8-oxo-78-dihydrodeoxyguanosine triphosphatase 8-oxo-78-dihydroguanosine triphosphatase 8-oxo-dGTPase Mth1 mutT human homolog (8-oxo-dGTPase) mutT human homolog 1 nucleoside diphosphate-linked moiety X motif 1 nucleoside diphosphate-linked moiety X-type motif 1 nudix (nucleoside diphosphate linked moiety X)-type motif 1 nudix hydrolase 1 Nudix motif 1 nudix-type motif 1 NUDT1; 7,8-dihydro-8-oxoguanine triphosphatase; 8-oxo-7,8-dihydro-2'-dGTPase; 8-oxo-7,8-dihydrodeoxyguanosine triphosphatase; 8-oxo-7,8-dihydroguanosine triphosphatase; 8-oxo-dGTPase; mutT human homolog (8-oxo-dGTPase); mutT human homolog 1; Nucleoside diphosphate-linked moiety X motif 1; nucleoside diphosphate-linked moiety X-type motif 1; nudix (nucleoside diphosphate linked moiety X)-type motif 1; Nudix motif 1; nudix-type motif 1
Gene Aliases: MTH1; NUDT1
UniProt ID: (Human) P36639, (Rat) P53369, (Mouse) P53368
Entrez Gene ID: (Human) 4521, (Rat) 117260, (Mouse) 17766
Molecular Function: hydrolase phosphorylase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.