Novus Biologicals
Manufacturer Code:NBP298486AF350
Catalog # NB098085
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human MTG1 fragment. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP GTP-binding protein 7 GTP-binding protein 7 (putative) GTPBP7 mitochondrial GTPase 1 mitochondrial GTPase 1 homolog (S. cerevisiae) RP11-108K14.2; GTP-binding protein 7; GTP-binding protein 7 (putative); Mitochondrial GTPase 1; mitochondrial GTPase 1 homolog; Mitochondrial ribosome-associated GTPase 1
Gene Aliases: GTP; GTPBP7; MTG1
UniProt ID: (Human) Q9BT17
Entrez Gene ID: (Human) 92170
Molecular Function: G-protein enzyme modulator hydrolase signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.