Proteintech
Manufacturer Code:137421AP150UL
Catalog # 50-172-8975
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MTG1 Fusion Protein Ag4681 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP binding protein 7 (putative); GTP GTP binding protein 7 GTPBP7 Mitochondrial GTPase 1 MTG1 RP11 108K14.2; GTP-binding protein 7; GTP-binding protein 7 (putative); Mitochondrial GTPase 1; mitochondrial GTPase 1 homolog; mitochondrial GTPase 1-like; Mitochondrial ribosome-associated GTPase 1
Gene Aliases: Gm169; GTP; GTPBP7; MTG1
UniProt ID: (Human) Q9BT17, (Mouse) Q8R2R6
Entrez Gene ID: (Human) 92170, (Rat) 100910751, (Mouse) 212508
Molecular Function: G-protein enzyme modulator hydrolase signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.