Invitrogen
Manufacturer Code:PA567032
Catalog # PIPA567032
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human MTF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPCl2; Metal regulatory transcription factor 2; metal response element-binding transcription factor 2; Metal-response element DNA-binding protein M96; Metal-response element-binding transcription factor 2; polycomb-like 2; Polycomb-like protein 2; putative DNA binding protein; TDRD19A dJ976O13.2 RP5-976O13.1 PCL2 M96 metal response element binding transcription factor 2; tudor domain containing 19A
Gene Aliases: dJ976O13.2; M96; MTF2; PCL2; TDRD19A
UniProt ID: (Human) Q9Y483
Entrez Gene ID: (Human) 22823
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.