OriGene
Manufacturer Code:TA805480
Catalog # 50-168-0718
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2F3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-272 of human MTF1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Metal regulatory transcription factor 1; metal-regulatory transcription factor 1; metal-responsive transcription factor 1; MRE-binding transcription factor; MRE-binding transcription factor-1; MTF-1 ZRF; Transcription factor MTF-1; zinc regulatory factor
Gene Aliases: MTF-1; MTF1; ZRF
UniProt ID: (Human) Q14872
Entrez Gene ID: (Human) 4520
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.