Novus Biologicals
Manufacturer Code:NBP275731
Catalog # NBP275731
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MTAP/1813 |
Immunogen | Recombinant human MTAP protein fragment (aa97-196) (exact sequence is proprietary) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse Monoclonal Antibody
Protein Aliases: 5'-methylthioadenosine phosphorylase; 5'-methylthioadenosine phosphorylase c86fus EC 2.4.2.28 methylthioadenosine phosphorylase MSAPMeSAdo phosphorylase MTA phosphorylase MTAPase S-methyl-5'-thioadenosine phosphorylase; epididymis luminal protein 249; MeSAdo phosphorylase; MTA phosphorylase; MTAP; MTAPase; S-methyl-5'-thioadenosine phosphorylase
Gene Aliases: BDMF; c86fus; DMSFH; DMSMFH; HEL-249; LGMBF; MSAP; MTAP
UniProt ID: (Human) Q13126
Entrez Gene ID: (Human) 4507
Molecular Function: phosphorylase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.