Invitrogen
Manufacturer Code:PA5106861
Catalog # PIPA5106861
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MTAP corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1300019I21Rik 5'-methylthioadenosine phosphorylase BDMF c86fus Cc1-6 DMSFH DMSMFH epididymis luminal protein 249 HEL-249 LGMBF MeSAdo phosphorylase methylthioadenosine phosphorylase MSAP MTA phosphorylase MTAP MTAPase S-methyl-5'-thioadenosine phosphorylase; 5'-methylthioadenosine phosphorylase; epididymis luminal protein 249; MeSAdo phosphorylase; MTA phosphorylase; MTAP; MTAPase; S-methyl-5'-thioadenosine phosphorylase
Gene Aliases: 1300019I21Rik; BDMF; c86fus; Cc1-6; DMSFH; DMSMFH; HEL-249; LGMBF; MSAP; MTAP
UniProt ID: (Human) Q13126, (Mouse) Q9CQ65
Entrez Gene ID: (Human) 4507, (Mouse) 66902, (Rat) 298227
Molecular Function:
phosphorylase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.