Invitrogen
Manufacturer Code:PA596107
Catalog # PIPA596107
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 40-330 of human MTA3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: metastasis associated 1 family, member 3; metastasis associated family, member 3; metastasis associated gene family, member 3; Metastasis-associated protein MTA3
Gene Aliases: 1110002J22Rik; KIAA1266; mKIAA1266; MTA3
UniProt ID: (Human) Q9BTC8, (Mouse) Q924K8
Entrez Gene ID: (Human) 57504, (Mouse) 116871
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
histone
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.