Invitrogen
Manufacturer Code:PA5100022
Catalog # PIPA5100022
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MTA3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110002J22Rik KIAA1266 metastasis associated 1 family member 3 metastasis associated 1 family member 3 metastasis associated 3 metastasis associated family member 3 metastasis associated gene family member 3 metastasis-associated protein MTA3 mKIAA1266 Mta3; metastasis associated 1 family, member 3; metastasis associated 3; metastasis associated family, member 3; metastasis associated gene family, member 3; Metastasis-associated protein MTA3
Gene Aliases: 1110002J22Rik; KIAA1266; mKIAA1266; MTA3
UniProt ID: (Human) Q9BTC8, (Mouse) Q924K8
Entrez Gene ID: (Human) 57504, (Mouse) 116871, (Rat) 100362346
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
histone
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.