Invitrogen
Manufacturer Code:PA582239
Catalog # PIPA582239
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human MTA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: metastais-associated gene family, member 2; metastasis -associated gene 1-like 1; metastasis associated 1 family, member 2; metastasis associated 1-like 1; metastasis associated gene family, member 2; Metastasis-associated 1-like 1; metastasis-associated gene family, member 2; metastasis-associated protein 2; Metastasis-associated protein MTA2; MTA1-L1 MTA1L1; MTA1-L1 protein; p53 target protein in deacetylase complex
Gene Aliases: AW550797; Mata1l1; mmta2; MTA1L1; MTA2; PID
UniProt ID: (Human) O94776, (Mouse) Q9R190
Entrez Gene ID: (Human) 9219, (Rat) 361724, (Mouse) 23942
Molecular Function: DNA binding protein chromatin/chromatin-binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.