Invitrogen
Manufacturer Code:PA599451
Catalog # PIPA599451
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MTA1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: metastasis associated 1 metastasis associated gene 1 protein metastasis-associated protein MTA1 MTA1 Zg29; metastasis associated gene 1 protein; Metastasis-associated protein MTA1
Gene Aliases: MTA1; Zg29
UniProt ID: (Human) Q13330, (Rat) Q62599
Entrez Gene ID: (Human) 9112, (Rat) 64520, (Mouse) 116870
Molecular Function: DNA binding protein chromatin/chromatin-binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.