Invitrogen
Manufacturer Code:PA578277
Catalog # PIPA578277
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human MTA1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: metastasis associated gene 1 protein; Metastasis-associated protein MTA1; MTA1 metastasis-associated protein MTA1 metastasis associated gene 1 protein metastasis associated 1
Gene Aliases: MTA1; Zg29
UniProt ID: (Human) Q13330, (Rat) Q62599
Entrez Gene ID: (Human) 9112, (Rat) 64520, (Mouse) 116870
Molecular Function: DNA binding protein chromatin/chromatin-binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.