Invitrogen
Manufacturer Code:PA131417
Catalog # PIPA131417
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide PGDVFYMPKE (Human). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: metastasis associated gene 1 protein; Metastasis-associated protein MTA1
Gene Aliases: MTA1
UniProt ID: (Human) Q13330
Entrez Gene ID: (Human) 9112
Molecular Function: DNA binding protein chromatin/chromatin-binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.