Novus Biologicals
Manufacturer Code:NB100823
Catalog # NB100823
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-PGDVFYMPKE corresponding to internal region (near C-Terminus) according to AAA78935.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: metastasis associated 1 metastasis associated gene 1 protein metastasis associated protein metastasis-associated protein MTA1; metastasis associated gene 1 protein; Metastasis-associated protein MTA1
Gene Aliases: MTA1
UniProt ID: (Human) Q13330
Entrez Gene ID: (Human) 9112
Molecular Function: DNA binding protein chromatin/chromatin-binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.