Proteintech
Manufacturer Code:121791AP150UL
Catalog # 50-172-7773
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MT3 Fusion Protein Ag2823 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GIF; GIF GIFB GRIF Growth inhibitory factor metallothionein 3 Metallothionein III MT 3 MT III MT3; GIFB; Growth inhibitory factor; metallothionein 3 (growth inhibitory factor (neurotrophic)); metallothionein III; Metallothionein-3; Metallothionein-III; MT-3; MT-III
Gene Aliases: GIF; GIFB; GRIF; Mt-3; MT3; ZnMT3
UniProt ID: (Human) P25713, (Rat) P37361, (Mouse) P28184
Entrez Gene ID: (Human) 4504, (Rat) 117038, (Mouse) 17751
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.