Invitrogen
Manufacturer Code:PA576652
Catalog # PIPA576652
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human MT2A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: metallothionein 2A; Metallothionein-2; Metallothionein-2 Metallothionein 2 MT-2 Metallothionein-2A Metallothionein-II MT-II MT2A CES1 MT2; Metallothionein-2A; Metallothionein-II; MT-2; MT-II
Gene Aliases: CES1; MT2; MT2A
UniProt ID: (Human) Q14823, (Rat) P04355
Entrez Gene ID: (Human) 4502, (Rat) 689415
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.