Proteintech
Manufacturer Code:145521AP150UL
Catalog # 50-172-9537
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MT1-MMP Fusion Protein Ag5965 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: matrix metallopeptidase 14 (membrane-inserted); matrix metalloproteinase 14 (membrane-inserted); matrix metalloproteinase 14 (membrane-inserted) alpha; matrix metalloproteinase 14 membrane-inserted; Matrix metalloproteinase 14 MMP 14 MMP X1 MMP14 MT MMP 1 MT1 MMP MT1MMP MT1-MMP MTMMP1; matrix metalloproteinase 14, membrane-inserted; matrix metalloproteinase 14a (membrane-inserted); Matrix metalloproteinase-14; membrane type 1 metalloprotease; membrane type 1-matrix metalloproteinase; Membrane type 1-MMP; Membrane-type matrix metalloproteinase 1; Membrane-type-1 matrix metalloproteinase; MMP-14; MMP-X1; MT-MMP; MT-MMP 1; MT-MMPalpha; MT1-MMP; MT1-MMP-alpha; MT1MMP; MTMMP1; type 1 matrix metalloprotease 14; zMT-MMP-alpha; zMT-MMPalpha
Gene Aliases: AI325305; cb386; MMP-14; MMP-X1; MMP14; MT-MMP; MT-MMP 1; MT-MMP-1; MT1-MMP; MT1MMP; Mtmmp; MTMMP1; sabe; sb:cb386; WNCHRS; wu:fb72b09
UniProt ID: (Human) P50281, (Mouse) P53690, (Rat) Q10739
Entrez Gene ID: (Human) 4323, (Mouse) 17387, (Rat) 81707, (Zebrafish) 373882
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.