R&D Systems
Manufacturer Code:MAB7917SP
Catalog # MAB7917SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 786607 |
Immunogen | <EM>E. coli</EM>-derived recombinant human MSX2 Ala2-Thr132 Accession # P35548 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CRS2 CRS2parietal foramina 1 FPP Homeobox protein Hox-8 homeobox protein MSX-2 Hox8 HOX8msh homeobox homolog 2 (Drosophila) MSH msh (Drosophila) homeo box homolog 2 msh homeo box 2 msh homeobox 2 msh homeobox homolog 2 PFM PFM1; Homeobox protein Hox-8; Homeobox protein MSX-2; msh homeo box 2; msh homeobox homolog 2
Gene Aliases: CRS2; FPP; HOX8; MSH; MSX2; PFM; PFM1
UniProt ID: (Human) P35548
Entrez Gene ID: (Human) 4488
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.