Invitrogen
Manufacturer Code:PA5106670
Catalog # PIPA5106670
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MSX2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BB122635 CRS2 FPP homeo box msh-like 2 homeobox protein Hox-8 Homeobox protein Hox-8-1 homeobox protein MSX-2 homeobox msh-like 2 Hox8 Hox-8 Hox8.1 Hox-8.1 MSH msh homeo box 2 msh homeo box homolog 2 msh homeobox 2 msh homeobox homolog 2 MSX2 Msx-2 PFM PFM1; homeo box, msh-like 2; Homeobox protein Hox-8; Homeobox protein Hox-8-1; Homeobox protein MSX-2; homeobox, msh-like 2; msh homeo box 2; msh homeobox homolog 2
Gene Aliases: BB122635; CRS2; FPP; Hox-8; Hox-8.1; HOX8; Hox8.1; MSH; Msx-2; MSX2; PFM; PFM1
UniProt ID: (Human) P35548, (Mouse) Q03358
Entrez Gene ID: (Human) 4488, (Mouse) 17702
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.