Novus Biologicals
Manufacturer Code:NBP185445
Catalog # NBP185445
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SSLPFSVEALMSDKKPPKEASPLPAESASAGATLRPLLLSGHGAREAHSPGPLVKPFETASVKSENSEDGAAWMQEPGRYSPPPRHTSPTTCTLRKHKTNRKP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CRS2parietal foramina 1 FPP Homeobox protein Hox-8 homeobox protein MSX-2 HOX8msh homeobox homolog 2 (Drosophila) MSH msh (Drosophila) homeo box homolog 2 msh homeo box 2 msh homeobox 2 msh homeobox homolog 2 PFM PFM1; Homeobox protein Hox-8; Homeobox protein MSX-2; msh homeo box 2; msh homeobox homolog 2
Gene Aliases: CRS2; FPP; HOX8; MSH; MSX2; PFM; PFM1
UniProt ID: (Human) P35548
Entrez Gene ID: (Human) 4488
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.