Abnova
Manufacturer Code:H00004488K
Catalog # 89-200-129
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human MSX2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human MSX2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: CRS2 FPP HOX8 MSH PFM PFM1; Homeobox protein Hox-8; Homeobox protein MSX-2; msh homeo box 2; msh homeobox homolog 2
Gene Aliases: CRS2; FPP; HOX8; MSH; MSX2; PFM; PFM1
UniProt ID: (Human) P35548
Entrez Gene ID: (Human) 4488
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.