R&D Systems
Manufacturer Code:MAB5045
Catalog # MAB5045
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 801601 |
Immunogen | <EM>E. coli</EM>-derived recombinant human MSX1 Met1-Thr165 Accession # P28360 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: homeobox 7; Homeobox protein Hox-7; Homeobox protein Hox-7 homeobox protein MSX-1 HOX7 HOX7homeobox 7 HYD1 HYD1msh homeobox homolog 1 (Drosophila) msh (Drosophila) homeo box homolog 1 (formerly homeo box 7) msh homeo box 1 msh homeobox 1 Msh homeobox 1-like protein msh homeobox homolog 1 OFC5 STHAG1; Homeobox protein MSX-1; msh homeo box 1; Msh homeobox 1-like protein; msh homeobox homolog 1
Gene Aliases: ECTD3; HOX7; HYD1; MSX1; STHAG1
UniProt ID: (Human) P28360
Entrez Gene ID: (Human) 4487
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.