Invitrogen
Manufacturer Code:PA535227
Catalog # PIPA535227
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 111-138 amino acids from the central region of human MSX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homeo box, msh-like 1; homeobox 7; Homeobox protein Hox-7; Homeobox protein MSX-1; homeobox, msh-like 1; Hox-7.1; msh homeo box 1; Msh homeobox 1-like protein; msh homeobox homolog 1; MSX1 HOX7 Homeobox protein MSX-1 Homeobox protein Hox-7 Msh homeobox 1-like protein; muscle-segment homeobox
Gene Aliases: AA675338; AI324650; ECTD3; Hox-7; HOX7; Hox7.1; HYD1; msh; MSX1; STHAG1
UniProt ID: (Human) P28360, (Mouse) P13297
Entrez Gene ID: (Human) 4487, (Mouse) 17701
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.