Novus Biologicals
Manufacturer Code:NB300808
Catalog # NB300808
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence TSLPLGVKVEDS-C corresponding to N-Terminus according to NP_002439.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: homeobox 7; Homeobox protein Hox-7; Homeobox protein Hox-7 homeobox protein MSX-1 HOX7homeobox 7 HYD1msh homeobox homolog 1 (Drosophila) msh (Drosophila) homeo box homolog 1 (formerly homeo box 7) msh homeo box 1 msh homeobox 1 Msh homeobox 1-like protein msh homeobox homolog 1 OFC5 STHAG1; Homeobox protein MSX-1; msh homeo box 1; Msh homeobox 1-like protein; msh homeobox homolog 1
Gene Aliases: ECTD3; HOX7; HYD1; MSX1; STHAG1
UniProt ID: (Human) P28360
Entrez Gene ID: (Human) 4487
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.