Abnova
Manufacturer Code:PAB6647
Catalog # 89-132-562
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human MSX1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of MSX1.
For Research Use Only
Protein Aliases: homeobox 7; Homeobox protein Hox-7; Homeobox protein MSX-1; HOX7; HYD1; msh homeo box 1; Msh homeobox 1-like protein; msh homeobox homolog 1
Gene Aliases: ECTD3; HOX7; HYD1; MSX1; STHAG1
UniProt ID: (Human) P28360
Entrez Gene ID: (Human) 4487
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.