Proteintech
Manufacturer Code:176781AP150UL
Catalog # 50-173-1717
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MSX1 Fusion Protein Ag11700 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homeo box, msh-like 1; homeobox 7; Homeobox protein Hox 7 Homeobox protein MSX 1 HOX7 HYD1 msh homeobox 1 Msh homeobox 1 like protein MSX1; Homeobox protein Hox-7; Homeobox protein MSX-1; homeobox, msh-like 1; Hox-7.1; msh homeo box 1; Msh homeobox 1-like protein; msh homeobox homolog 1; muscle-segment homeobox
Gene Aliases: AA675338; AI324650; ECTD3; Hox-7; HOX7; Hox7.1; HYD1; msh; MSX1; STHAG1
UniProt ID: (Human) P28360, (Mouse) P13297
Entrez Gene ID: (Human) 4487, (Mouse) 17701, (Rat) 81710
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.