Invitrogen
Manufacturer Code:PA5115278
Catalog # PIPA5115278
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GDF8 / Myostatin corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cmpt compact GDF8 GDF-8 growth differentiation factor 8 growth/differentiation factor 8 H-Myo-Propeptide H-Myostatin MSLHP MSTN myostatin; compact; GDF-8; growth differentiation factor 8; Growth/differentiation factor 8; Myostatin
Gene Aliases: Cmpt; GDF8; MSLHP; MSTN
UniProt ID: (Human) O14793, (Rat) O35312, (Mouse) O08689
Entrez Gene ID: (Human) 2660, (Rat) 29152, (Mouse) 17700
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.