Invitrogen
Manufacturer Code:PA522015
Catalog # PIPA522015
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 324 and 418 of STK4 (Uniprot IDNo.Q13043) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: kinase responsive to stress 2; KRS2 MST1 YSK3; Mammalian STE20-like protein kinase 1; mammalian sterile 20-like 1; MST-1; MST1/C; MST1/N; Serine/threonine-protein kinase 4; Serine/threonine-protein kinase 4 18kDa subunit; Serine/threonine-protein kinase 4 37kDa subunit; Serine/threonine-protein kinase Krs-2; STE20-like kinase MST1; sterile 20-like kinase 1; Yeast Sps1/Ste20-related kinase 3
Gene Aliases: AI447339; AU020804; Kas-2; KRS2; MST1; STK4; YSK3
UniProt ID: (Human) Q13043, (Mouse) Q9JI11
Entrez Gene ID: (Human) 6789, (Mouse) 58231
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.