Proteintech
Manufacturer Code:142511AP150UL
Catalog # 50-172-9318
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MSRB3 Fusion Protein Ag5524 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: methionine sulfoxide reductase B3; Methionine-R-sulfoxide reductase B3; Methionine-R-sulfoxide reductase B3, mitochondrial; MSRB3
Gene Aliases: D430026P16Rik; DFNB74; I-8-23; MsrB2; MSRB3; UNQ1965/PRO4487
UniProt ID: (Human) Q8IXL7, (Mouse) Q8BU85
Entrez Gene ID: (Human) 253827, (Mouse) 320183, (Rat) 680036
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.