Novus Biologicals
Manufacturer Code:NBP297958F
Catalog # NB049989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human MSRB2 fragment. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CBS1 CBS-1methionine-r-sulfoxide reductase B2 variant 3 CGI-131 EC 1.8.4 EC 1.8.4.- EC 1.8.4.12 methionine sulfoxide reductase B methionine sulfoxide reductase B2 methionine-r-sulfoxide reductase B2 variant 1 methionine-R-sulfoxide reductase B2 mitochondrial MGC26104 MSRB MsrB2 PILB pilin-like transcription factor; Methionine-R-sulfoxide reductase B2, mitochondrial; MsrB2; pilin-like transcription factor
Gene Aliases: CBS-1; CBS1; CGI-131; MSRB; MSRB2; PILB
UniProt ID: (Human) Q9Y3D2
Entrez Gene ID: (Human) 22921
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.