Proteintech
Manufacturer Code:176291AP150UL
Catalog # 50-173-1694
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MSRB2 Fusion Protein Ag11881 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CBS 1 CBS1 CGI 131 MSRB MSRB2 PILB; Methionine-R-sulfoxide reductase B2, mitochondrial; MsrB2; pilin-like transcription factor
Gene Aliases: 2310050L06Rik; CBS-1; CBS1; CGI-131; Mrsb; MSRB; MSRB2; PILB
UniProt ID: (Human) Q9Y3D2, (Rat) Q4FZX5, (Mouse) Q78J03
Entrez Gene ID: (Human) 22921, (Rat) 361286, (Mouse) 76467
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.