Invitrogen
Manufacturer Code:PA577009
Catalog # PIPA577009
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human MSRB1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Methionine-R-sulfoxide reductase B1; Methionine-R-sulfoxide reductase B1 MsrB1 Selenoprotein X SelX MSRB1 SEPX1 HSPC270; MsrB1; selenoprotein R; Selenoprotein X; selenoprotein X, 1; SelX
Gene Aliases: HSPC270; MSRB1; SELR; SELX; SepR; SEPX1
UniProt ID: (Human) Q9NZV6
Entrez Gene ID: (Human) 51734
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.