Invitrogen
Manufacturer Code:PA514206
Catalog # PIPA514206
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 34-63 amino acids from the N-terminal region of human MSRA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cytosolic methionine-S-sulfoxide reductase; Mitochondrial peptide methionine sulfoxide reductase; peptide met (O) reductase; Peptide Met(O) reductase; peptide methionine sulfoxide reductase; Peptide methionine sulfoxide reductase Protein-methionine-S-oxide reductase PMSR Peptide-methionine (S)-S-oxide reductase Peptide Met(O) reductase MSRA; Peptide-methionine (S)-S-oxide reductase; PMSR; Protein-methionine-S-oxide reductase
Gene Aliases: 2310045J23Rik; 6530413P12Rik; MSR-A; MSRA; PMSR
UniProt ID: (Human) Q9UJ68, (Mouse) Q9D6Y7
Entrez Gene ID: (Human) 4482, (Mouse) 110265
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.