Proteintech
Manufacturer Code:145471AP150UL
Catalog # 50-172-9534
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MSRA Fusion Protein Ag6053 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cytosolic methionine-S-sulfoxide reductase; Mitochondrial peptide methionine sulfoxide reductase; MSRA Peptide Met(O) reductase PMSR; peptide met (O) reductase; Peptide Met(O) reductase; peptide methionine sulfoxide reductase; Peptide-methionine (S)-S-oxide reductase; PMSR; Protein-methionine-S-oxide reductase
Gene Aliases: 2310045J23Rik; 6530413P12Rik; MSR-A; MSRA; PMSR
UniProt ID: (Human) Q9UJ68, (Rat) Q923M1, (Mouse) Q9D6Y7
Entrez Gene ID: (Human) 4482, (Rat) 29447, (Mouse) 110265
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.