R&D Systems
Manufacturer Code:AF1947SP
Catalog # AF1947SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Phosphopeptide containing human MSP R/Ron Y1238/Y1239 sites |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-met-related tyrosine kinase; CD136; CD136 CD136 antigen CDw136c-met-related tyrosine kinase EC 2.7.10 EC 2.7.10.1 macrophage stimulating 1 receptor (c-met-related tyrosine kinase) MSP R MSP receptor MST1R p185-Ron Protein-tyrosine kinase 8 PTK8 PTK8 protein tyrosine kinase 8 Ron RONmacrophage-stimulating protein receptor soluble RON variant 1 soluble RON variant 2 soluble RON variant 3 soluble RON variant 4; CDw136; Macrophage-stimulating protein receptor; Macrophage-stimulating protein receptor alpha chain; Macrophage-stimulating protein receptor beta chain; MSP receptor; p185-Ron; Protein-tyrosine kinase 8; PTK8 protein tyrosine kinase 8
Gene Aliases: CD136; CDw136; MST1R; PTK8; RON
UniProt ID: (Human) Q04912
Entrez Gene ID: (Human) 4486
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.