R&D Systems
Manufacturer Code:FAB6647P
Catalog # FAB6647P
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human MSP R/Ron Glu25-Ser956 Accession # Q04912 |
Conjugate | R-PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: c-met-related tyrosine kinase; CD136; CD136 CD136 antigen CDw136c-met-related tyrosine kinase EC 2.7.10 EC 2.7.10.1 macrophage stimulating 1 receptor (c-met-related tyrosine kinase) MSP R MSP receptor MST1R p185-Ron Protein-tyrosine kinase 8 PTK8 PTK8 protein tyrosine kinase 8 Ron RONmacrophage-stimulating protein receptor soluble RON variant 1 soluble RON variant 2 soluble RON variant 3 soluble RON variant 4; CDw136; Macrophage-stimulating protein receptor; Macrophage-stimulating protein receptor alpha chain; Macrophage-stimulating protein receptor beta chain; MSP receptor; p185-Ron; Protein-tyrosine kinase 8; PTK8 protein tyrosine kinase 8
Gene Aliases: CD136; CDw136; MST1R; PTK8; RON
UniProt ID: (Human) Q04912
Entrez Gene ID: (Human) 4486
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.