R&D Systems
Manufacturer Code:AF189
Catalog # AF189
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Phosphopeptide containing human and mouse MSK2/RSKB S196 site |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after opening. 6 months -20 to -70 degreesC under sterile conditions |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 90 kDa ribosomal protein S6 kinase 4; mitogen- and stress-activated protein kinase 2; Nuclear mitogen- and stress-activated protein kinase 2; Ribosomal protein kinase B; ribosomal protein S6 kinase 90kDa polypeptide 4 RPS6KA4 RSKB; ribosomal protein S6 kinase alpha 4; Ribosomal protein S6 kinase alpha-4; ribosomal protein S6 kinase, 90kDa, polypeptide 4; RSKB; S6K-alpha-4
Gene Aliases: MSK2; RPS6KA4; RSK-B; S6K-alpha-4
UniProt ID: (Human) O75676
Entrez Gene ID: (Human) 8986
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.