Invitrogen
Manufacturer Code:PA5115594
Catalog # PIPA5115594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MSK2 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110069D02Rik 90 kDa ribosomal protein S6 kinase 4 90kDa AI848992 Ccdc88b coiled-coil domain containing 88B EC 2.7.11.1 kinase MSK2 KS6A4 mitogen- and stress-activated protein kinase 2 mitogen- and stress-activated protein kinase-2 mMSK2 MSK2 Nuclear mitogen- and stress-activated protein kinase 2 ribosomal protein kinase B ribosomal protein S6 kinase A4 ribosomal protein S6 kinase alpha 4 ribosomal protein S6 kinase alpha-4 ribosomal protein S6 kinase 90kD polypeptide 4 ribosomal protein S6 kinase 90kDa polypeptide 4 ribosomal protein S6 kinase polypeptide 4 RLSK Rps6ka4 RSKB RSK-B RSK-like protein kinase S6K-alpha-4; 90 kDa ribosomal protein S6 kinase 4; mitogen- and stress-activated protein kinase 2; mitogen- and stress-activated protein kinase-2; Nuclear mitogen- and stress-activated protein kinase 2; Ribosomal protein kinase B; ribosomal protein S6 kinase alpha 4; Ribosomal protein S6 kinase alpha-4; ribosomal protein S6 kinase, 90kD, polypeptide 4; ribosomal protein S6 kinase, 90kDa, polypeptide 4; RLSK; RSK-like protein kinase; RSKB; S6K-alpha-4
Gene Aliases: 1110069D02Rik; 90kDa; AI848992; mMSK2; MSK2; RPS6KA4; RSK-B; S6K-alpha-4
UniProt ID: (Human) O75676, (Mouse) Q9Z2B9
Entrez Gene ID: (Human) 8986, (Mouse) 56613
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.