R&D Systems
Manufacturer Code:AF2518SP
Catalog # AF2518SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human MSK1 Met1-Ala802 Accession # O75582 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 90 kDa ribosomal protein S6 kinase 5; EC 2.7.11 MGC191190 kDa ribosomal protein S6 kinase 5 MSK1EC 2.7.11.1 MSPK1 Nuclear mitogen- and stress-activated protein kinase 1 ribosomal protein S6 kinase alpha-5 ribosomal protein S6 kinase 90kD polypeptide 5 ribosomal protein S6 kinase 90kDa polypeptide 5 RLPK RPS6KA5 RSKL RSK-like protein kinase S6K-alpha-5; Nuclear mitogen- and stress-activated protein kinase 1; Ribosomal protein S6 kinase alpha-5; ribosomal protein S6 kinase, 90kDa, polypeptide 5; RSK-like protein kinase; RSKL; S6K-alpha-5
Gene Aliases: MSK1; MSPK1; RLPK; RPS6KA5
UniProt ID: (Human) O75582
Entrez Gene ID: (Human) 9252
Molecular Function: annexin calcium-binding protein calmodulin cytoskeletal protein intracellular calcium-sensing protein kinase microtubule family cytoskeletal protein non-motor microtubule binding protein non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.