Invitrogen
Manufacturer Code:PA538371
Catalog # PIPA538371
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic non-phosphopeptide derived from human MSK1 around the phosphorylation site of Thr581 (L-K-TP-P-C) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 90 kDa ribosomal protein S6 kinase 5; Nuclear mitogen- and stress-activated protein kinase 1; Ribosomal protein S6 kinase alpha-5; Ribosomal protein S6 kinase alpha-5 S6K-alpha-5 90 kDa ribosomal protein S6 kinase 5 Nuclear mitogen- and stress-activated protein kinase 1 RSK-like protein kinase RSKL RPS6KA5 MSK1; ribosomal protein S6 kinase, 90kDa, polypeptide 5; RLSK; RSK-like protein kinase; RSKL; S6K-alpha-5
Gene Aliases: 3110005L17Rik; 6330404E13Rik; AI854034; MSK1; MSPK1; RLPK; RPS6KA5
UniProt ID: (Human) O75582, (Mouse) Q8C050
Entrez Gene ID: (Human) 9252, (Mouse) 73086
Molecular Function: annexin calcium-binding protein calmodulin cytoskeletal protein intracellular calcium-sensing protein kinase microtubule family cytoskeletal protein non-motor microtubule binding protein non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.