Invitrogen
Manufacturer Code:711684
Catalog # PI711684
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Immunogen | Peptides corresponding to human MSK1 &lsqb1) aa742-aa758 2) aa773-aa792] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: 90 kDa ribosomal protein S6 kinase 5; MSK1; Nuclear mitogen- and stress-activated protein kinase 1; Ribosomal protein S6 kinase alpha-5; ribosomal protein S6 kinase, 90kDa, polypeptide 5; RSK-like protein kinase; RSKL; S6K-alpha-5
Gene Aliases: MSK1; MSPK1; RLPK; RPS6KA5
UniProt ID: (Human) O75582
Entrez Gene ID: (Human) 9252
Molecular Function:
annexin
calcium-binding protein
calmodulin
cytoskeletal protein
intracellular calcium-sensing protein
kinase
microtubule family cytoskeletal protein
non-motor microtubule binding protein
non-receptor serine/threonine protein kinase
protein kinase
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.