Proteintech
Manufacturer Code:107701AP150UL
Catalog # 50-172-6582
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MSI2 Fusion Protein Ag1213 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MSI2 MSI2H Musashi 2 musashi homolog 2 (Drosophila); Musashi homolog 2; musashi RNA-binding protein 2; Musashi-2; Musashi2; RNA-binding protein Musashi homolog 2
Gene Aliases: 1700105C15Rik; AI451722; AI563628; AW489193; MSI2; MSI2H; RGD1560397
UniProt ID: (Human) Q96DH6, (Mouse) Q920Q6
Entrez Gene ID: (Human) 124540, (Mouse) 76626, (Rat) 360596
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.